Learn More
STEMCELL Technologies Human Recombinant bFGF, ACF
Product Code. 17179641
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Product Code. 17179641
Brand: STEMCELL Technologies 78134
Basic fibroblast growth factor, animal component-free
  • Basic fibroblast growth factor (bFGF) is a prototypic member of the fibroblast growth factor family
  • Cytokines in the FGF family possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including cell proliferation, differentiation, survival, and apoptosis
  • bFGF has the β-trefoil structure, binds to the four FGF receptor (FGFR) family members, and activates JAK/STAT, PI3K, ERK1/2, and other receptor tyrosine kinase (RTK) signaling pathways
  • It supports the maintenance of undifferentiated human pluripotent stem cells, stimulates human pluripotent stem cells to form neural rosettes, and improves proliferation of human mesenchymal stem cells and enhances chondrogenic differentiation
Specifications
Name Basic fibroblast growth factor, FGF-β, FGF2, Fibroblast growth factor-basic, HBGF-2
Quantity 10 μg
Cell Type Brain Tumor Stem Cells; Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Pluripotent Stem Cells; Prostate Cells
Purity or Quality Grade ≥95%