missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant BDNF
Description
- Brain-derived neurotrophic factor (BDNF), like nerve growth factor (NGF), neurotrophin-3 (NT-3), and neurotrophin-4 (NT-4), is a member of the NGF family of neurotrophins, which are required for the differentiation and survival of specific neuronal subpopulations in both the central and the peripheral nervous systems
- BDNF binds with high affinity to the tropomyosin receptor kinase B (TrkB), and activates AKT and ERK pathways
- It is expressed in the hippocampus, cortex, and synapses of the basal forebrain
- BDNF acts as a survival factor for human embryonic stem cells when plated on either feeder cells or Corning™ Matrigel™
Specifications
Specifications
| Name | Abrineurin, ANON2, BULN2, Neurotrophin, MGC34632 |
| Quantity | 10 μg |
| Cell Type | Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons |
| Purity or Quality Grade | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?