missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant BAFF
Click to view available options
Quantity:
10 μg
1000 μg
50 μg
Unit Size:
1000µg
10µg
50µg
Description
- B cell activating factor (BAFF) is part of the tumor necrosis factor (TNF) superfamily and is expressed by monocytes, macrophages, dendritic cells, and neutrophils (Mackay & Browning; Scapini et al.) It binds to 3 receptors: BAFF-R, TACI, and BCMA, all of which are expressed by B cells at various times in their development (Liu & Davidson)
- BAFF-R signaling activates the NF-kB signaling pathway, and promotes B cell survival, Ig class switching, and antigen presentation (Bossen & Schneider; Liu & Davidson; Naradikian et al.) BAFF also has functions in other cell types: it promotes monocyte survival and activation, proinflammatory cytokine secretion, and differentiation into macrophages (Chang et al.), and co-stimulates anti-CD3-mediated activation of human T cells (Huard et al.; Mackay & Leung)
Specifications
Specifications
| Name | BLyS, TALL1, TNFSF13B |
| Quantity | 50 μg |
| Cell Type | B Cells |
| Purity or Quality Grade | >95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction