missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant Activin A, ACF
Description
- Activin A is a member of the transforming growth factor beta (TGF-β) family of proteins produced by many cell types throughout development (Gurdon et al.) It is a disulfide-linked homodimer (two beta-A chains) that binds to heteromeric complexes of a type I (Act RI-A and Act RI-B) and a type II (Act RII-A and Act RII-B) serine-threonine kinase receptor (Attisano et al.) Activins primarily signal through SMAD2/3 proteins to regulate a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, and metabolism (McDowell et al.) Activin A maintains the undifferentiated state of human embryonic stem cells (James et al.; Xiao et al.) and also facilitates differentiation of human embryonic stem cells into definitive endoderm (D'Amour et al.) This product is animal component-free
Specifications
Specifications
| Name | Activin beta-A chain, EDF, Erythroid differentiation protein, FRP, FSH-releasing protein, INHBA, Inhibin betaA chain, Inhibin beta-1 |
| Quantity | 10 μg |
| Cell Type | Endoderm, PSC-Derived; Mesoderm, PSC-Derived; Other; Pluripotent Stem Cells |
| Purity or Quality Grade | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?