missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RECK Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1734
This item is not returnable.
View return policy
Description
RECK Monoclonal specifically detects RECK in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| RECK | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| O95980 | |
| RECK | |
| Mouse myeloma cell line NS0-derived recombinant human RECK Ala29-Val934 Accession # O95980 | |
| 500 μg | |
| Primary | |
| Detects human RECK in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 198212 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hRECK, membrane-anchored glycoprotein (metastasis and invasion), reversion-inducing-cysteine-rich protein with kazal motifs, ST15, ST15reversion-inducing cysteine-rich protein with Kazal motifs, suppression of tumorigenicity 15 (reversion-inducing-cysteine-rich protein withkazal motifs), suppression of tumorigenicity 5 (reversion-inducing-cysteine-rich protein withkazal motifs), Suppressor of tumorigenicity 15 protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8434 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction