missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RDH13 (aa 220-291) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Packungsgröße:
100µL
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59818 (PA5-59818. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Does not exhibit retinol dehydrogenase (RDH) activity in vitro.
Spezifikation
Spezifikation
| Accession Number | Q8NBN7 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 112724 |
| Name | Human RDH13 (aa 220-291) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 8430425D21Rik; PSEC0082; Rdh13; Retinol dehydrogenase 13; retinol dehydrogenase 13 (all-trans and 9-cis); retinol dehydrogenase 13 (all-trans/9-cis); SDR7C3; Short chain dehydrogenase/reductase family 7 C member 3; short chain dehydrogenase/reductase family 7 C, member 3; UNQ736/PRO1430 |
| Common Name | RDH13 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur