missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RCN3 (aa 94-162) Control Fragment Recombinant Protein

Product Code. 30182100
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182100

Brand: Invitrogen™ RP99107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60252 (PA5-60252. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Probable molecular chaperone assisting protein biosynthesis and transport in the endoplasmic reticulum (PubMed:16433634, PubMed:28939891). Required for the proper biosynthesis and transport of pulmonary surfactant-associated protein A/SP-A, pulmonary surfactant-associated protein D/SP-D and the lipid transporter ABCA3. By regulating both the proper expression and the degradation through the endoplasmic reticulum-associated protein degradation pathway of these proteins plays a crucial role in pulmonary surfactant homeostasis. Has an anti-fibrotic activity by negatively regulating the secretion of type I and type III collagens (PubMed:28939891). This calcium-binding protein also transiently associates with immature PCSK6 and regulates its secretion (PubMed:16433634). [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96D15
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57333
Name Human RCN3 (aa 94-162) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6030455P07Rik; D530026G20Rik; D7Ertd671e; EF-hand calcium-binding protein RLP49; Rcn3; reticulocabin; reticulocalbin 3; reticulocalbin 3, EF-hand calcium binding domain; reticulocalbin-3; RLP49; UNQ239/PRO272
Common Name RCN3
Gene Symbol RCN3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DGWVSLAELRAWIAHTQQRHIRDSVSAAWDTYDTDRDGRVGWEELRNATYGHYAPGEEFHDVEDAETYK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.