missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human RBX1 Antibody, Novus Biologicals™
GREENER_CHOICE

Mouse Monoclonal Antibody

£195.00 - £469.00

Specifications

Antigen RBX1
Clone 1076128
Dilution Western Blot 2 μg/mL, Simple Western 100 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL
Applications Immunohistochemistry, Western Blot, Simple Western, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18754484
View Documents
Novus Biologicals™
MAB11499-SP
25 μg
£195.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18764534
GREENER_CHOICE
View Documents
Novus Biologicals™
MAB11499-100
100 μg
£469.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RBX1 Monoclonal antibody specifically detects RBX1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Simple Western,Immunocytochemistry
Specifications

Specifications

RBX1
Western Blot 2 μg/mL, Simple Western 100 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
BA554C12.1, EC 6.3.2.-, FLJ60363, MGC13357, Protein ZYP, Rbx1, Regulator of cullins 1, Ring Box 1, RING box protein 1, RING finger protein 75, ring-box 1, ring-box 1, E3 ubiquitin protein ligase, RING-box protein 1, RNF75, RNF75MGC1481, ROC1E3 ubiquitin-protein ligase RBX1, ZYP protein
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human CUL1/RBX1 Complex, Met1-His108, Accession # P62877
Primary
Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
1076128
Immunohistochemistry, Western Blot, Simple Western, Immunocytochemistry
Unconjugated
Mouse
Human
P62877
9978
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.