missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RBMXL2 (aa 298-320) Control Fragment Recombinant Protein

Product Code. 30201361
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201361

Brand: Invitrogen™ RP100851

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62497 (PA5-62497. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. RBMXL2 has two RRM domains that bind RNAs. RBMXL2 has similarity to HNRPG and RBMY proteins and it is suggested to replace HNRPG protein function during meiotic prophase or act as a germ cell-specific splicing regulator. It primarily localizes to the nuclei of meiotic spermatocytes. This gene is a candidate for autosomal male infertility.This gene belongs to the HNRPG subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75526
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 27288
Name Human RBMXL2 (aa 298-320) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700012H05Rik; Heterogeneous nuclear ribonucleoprotein G; heterogeneous nuclear ribonucleoprotein G T; hnRNP G; hnRNP G-T; HNRNPGT; Hnrnpg-t; HNRPG; HNRPGT; LOC617969 protein; NEWGENE_1598286; RBMX; RBMX like 2; Rbmxl2; RNA binding motif protein, X-linked like 2; RNA binding motif protein, X-linked-like 2; RNA-binding motif protein, x chromosome; RNA-binding motif protein, x chromosome, N-terminally processed; RNA-binding motif protein, X-linked-like-2; testes specific heterogenous nuclear ribonucleoprotein G T; testes-specific heterogenous nuclear ribonucleoprotein G-T; testis-specific heterogeneous nuclear ribonucleoprotein G-T
Common Name RBMXL2
Gene Symbol RBMXL2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TPPSYGGGGRYEEYRGYSPDAYS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.