missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RBBP9 (aa 117-186) Control Fragment Recombinant Protein
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
RBBP9 may play a role in the transformation process due to its capacity to confer resistance to the growth-inhibitory effects of TGF-beta1 through interaction with retinoblastoma and the subsequent displacement of E2F-1.The protein encoded by this gene is a retinoblastoma binding protein that may play a role in the regulation of cell proliferation and differentiation. Two alternatively spliced transcript variants of this gene with identical predicted protein products have been reported, one of which is a nonsense-mediated decay candidate.
Spezifikation
Spezifikation
| Accession Number | O75884 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 10741 |
| Name | Human RBBP9 (aa 117-186) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | B5T overexpressed gene protein; B5T-overexpressed gene protein; BOG; im:7247963; im:7247963 protein; MGC9236; Protein BOG; Putative hydrolase RBBP9; RB binding protein 9, serine hydrolase; RBBP10; RBBP-10; Rbbp9; RBBP-9; retinoblastoma binding protein 9; retinoblastoma-binding protein 10; Retinoblastoma-binding protein 9; retinoma-binding protein 9; serine hydrolase RBBP9; serine hydrolase RBBP9; putative hydrolase RBBP9; si:ch211-258l4.7; zgc:152807 |
| Common Name | RBBP9 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur