Learn More
Invitrogen™ Human RbAp48 (aa 1-14) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP084
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Human RbAp48 (residues 1-14) Synthetic Peptide, PEP-084, from Invitrogen.
PEP-084 is a 14 amino acid synthetic peptide whose sequence corresponds to residues 1-14 of human RbAp48 with a carboxy-terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): M1-A-D-K-F-A-A-F-D-D-A-V-E-E14-CThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and endogenous RbAp48 catalog # PA1-869. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
5928 | |
purified | |
Human RbAp48 (aa 1-14) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q09028 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
RBBP4 | |
No preservative | |
50 μg | |
M1 - A - D - K - F - A - A - F - D - D - A - V - E - E14 - C |