missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat SUSD2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB90563-SP
This item is not returnable.
View return policy
Description
SUSD2 Monoclonal specifically detects SUSD2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| SUSD2 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2 | |
| HEK293 human embryonic kidney cell line-derived human SUSD2 Met1-Ala785 Accession # Q9UGT4 | |
| 25 μg | |
| Primary | |
| Detects human SUSD2 in direct ELISAs and Western blots. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Flow Cytometry | |
| 1279D | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells | |
| Q9UGT4 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 56241 | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction