missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat PTK7/CCK4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4499-SP
This item is not returnable.
View return policy
Description
PTK7/CCK4 Monoclonal specifically detects PTK7/CCK4 in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| PTK7/CCK4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q13308 | |
| PTK7 | |
| E. coli-derived recombinant human PTK7/CCK4 Ala31-Ser199 Accession # Q13308 | |
| 25 μg | |
| Primary | |
| Detects human and rat PTK7/CCK4 in Western blots. | |
| Human, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 525222 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CCK4, CCK-4, CCK4Colon carcinoma kinase 4, colon carcinoma kinase-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, PTK7 protein tyrosine kinase 7, Tyrosine-protein kinase-like 7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5754 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction