missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat PKC beta 1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3759
This item is not returnable.
View return policy
Description
PKC beta 1 Polyclonal specifically detects PKC beta 1 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| PKC beta 1 | |
| Polyclonal | |
| Unconjugated | |
| P05771 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5579 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| PRKCB | |
| E. coli-derived recombinant human PKC beta 1 Lys607-Val671 Accession # P05771 | |
| 100 μg | |
| Primary | |
| Detects human and rat PKC beta 1 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human PKC delta is observed. | |
| Human, Rat | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu