missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat Osteocalcin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 24 publications
Brand: R&D Systems MAB1419-SP
This item is not returnable.
View return policy
Description
Osteocalcin Monoclonal specifically detects Osteocalcin in Human, Rat samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| Osteocalcin | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| P02818 | |
| BGLAP | |
| Human Osteocalcin synthetic peptide YLYQWLGAPVPYPDPLEPRREVCELNPDCDELADHIGFQEAYRRFYGPV Accession # P02818 | |
| 25 μg | |
| Primary | |
| Detects human Osteocalcin in direct ELISAs. | |
| Human, Rat | |
| Purified |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 190125 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BGLAP, BGP, bone gamma-carboxyglutamate (gla) protein, bone gamma-carboxyglutamate (gla) protein (osteocalcin), Bone Gla protein, Gamma-carboxyglutamic acid-containing protein, OC, OCN, osteocalcin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 632 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction