Learn More
Human/Rat Neuroligin 2/NLGN2 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems AF5645-SP
Product Code. 18709338
  • £168.00 / 25µg
Description
Neuroligin 2/NLGN2 Polyclonal specifically detects Neuroligin 2/NLGN2 in Human, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Neuroligin 2/NLGN2
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
EC 3.1.1, EC 3.1.1.1, KIAA1366neuroligin-2, neuroligin 2, NLGN2
Goat
Affinity Purified
RUO
57555
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 1-3 ug/mL
EAW90197
NLGN2
Mouse myeloma cell line NS0-derived recombinant human Neuroligin 2/NLGN2 Gln15-Ser660 Accession # EAW90197
25 μg
Primary
Detects human and rat Neuroligin 2/NLGN2 in direct ELISAs and Western blots.
Human, Rat