missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat CCR4 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 17 publications
£199.00 - £476.00
Specifications
| Antigen | CCR4 |
|---|---|
| Clone | 205410 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18789237
![]() |
R&D Systems
FAB1567P-025 |
25 Tests |
£199.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18759266
![]() |
R&D Systems
FAB1567P-100 |
100 Tests |
£476.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR4 Monoclonal specifically detects CCR4 in Human, Rat samples. It is validated for Flow Cytometry.Specifications
| CCR4 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Human, Rat | |
| C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
| CCR4 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Stains human CCR4-transfected cell lines but not the corresponding parent cell lines. Also detects human CCR4 on human PBLs and platelets and rat CCR4 on rat splenocytes. |
| 205410 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| P51679 | |
| 1233 | |
| Human CCR4 transfectants Met1-Leu360 Accession # P51679 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title