missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human/Rat BDNF ELISA Kit

Product Code. 16861506 Shop All Proteintech Products
Change view
Click to view available options
Target Species:
Human, Rat
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Target Species Unit Size
16861506 Human, Rat 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16861506 Supplier Proteintech™ Supplier No. KE0009696T
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

KE00096 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The BDNF ELISA kit is to be used to detect and quantify protein levels of endogenous BDNF.

  • The assay recognizes human BDNF. An antibody specific for BDNF has been pre-coated onto the microwells. The BDNF protein in samples is captured by the coated antibody after incubation.
  • Following extensive washing, another antibody of biotinylated specific for human BDNF is added to detect the captured human BDNF protein.
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
  • The brain-derived neurotrophic factor (BDNF), as a member of the neurotrophin family of growth factors, is broadly expressed in the central nervous system.
  • BDNF is involved in cerebral morphogenesis, synaptic plasticity and learning, mental health, degenerative central nervous system (CNS) diseases, and energy metabolism.
  • BDNF has recently been identified as one of the four major pharmaceutical targets in neuropsychiatric diseases, including depression and bipolar disorder.
  • In addition, BDNF is increasingly considered as a potential therapeutic agent for neurodevelopmental disorders such as Rett syndrome.

Specifications

Accession Number NP_001137277
Assay Range 12.5 to 800 pg/mL
Assay Sensitivity 0.1 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human, Rat
Gene ID (Entrez) 627
Host Species Human/Rat
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID P23560/P23363
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human/Rat BDNF ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.