Learn More
Human/Rat Annexin V Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB399-100
 View more versions of this product
Product Code. 18755059
  • £395.00 / 100µg
Description
Annexin V Monoclonal specifically detects Annexin V in Human, Rat samples. It is validated for Western Blot, Simple Western, Knockout Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Annexin V
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, ANXA5, ANXV, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha
E. coli-derived recombinant human Annexin V Met1-Asp320 Accession # P08758
100 μg
Primary
Detects human Annexin V in direct ELISAs and Western blots. Detects rat Annexin V in Simple Westerns TM .
Human, Rat
Purified
Western Blot
39103
Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL
P08758
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
308
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only (RUO)