missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat Angiogenin Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF265
This item is not returnable.
View return policy
Description
Angiogenin Polyclonal specifically detects Angiogenin in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Angiogenin | |
| Polyclonal | |
| Unconjugated | |
| Q53X86 | |
| ANG | |
| E. coli-derived recombinant human Angiogenin Gln25-Pro147 Accession # Q53X86 | |
| 100 μg | |
| Primary | |
| Detects human Angiogenin in direct ELISAs and Western blots. | |
| Human, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 1-15 ug/mL | |
| ALS9, ANG, angiogenin, angiogenin, ribonuclease, RNase A family, 5, EC 3.1.27, EC 3.1.27.-, epididymis luminal protein 168, HEL168, MGC22466, MGC71966, Ribonuclease 5, RNase 5, RNASE5RNASE4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 283 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido