missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RASSF8 (aa 149-233) Control Fragment Recombinant Protein

Product Code. 30199725
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199725

Brand: Invitrogen™ RP102815

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58277 (PA5-58277. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

RASSF8 encodes a member of the Ras-association domain family (RASSF) of tumor suppressor proteins. This gene is essential for maintaining adherens junction function in epithelial cells and has a role in epithelial cell migration. It is a lung tumor suppressor gene candidate. A chromosomal translocation t(12;22)(p11.2;q13.3) leading to the fusion of this gene and the FBLN1 gene is found in a complex type of synpolydactyly. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Spécification

Accession Number Q8NHQ8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11228
Name Human RASSF8 (aa 149-233) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5133400D11Rik; AA815821; AW123240; C12orf2; Carcinoma-associated protein HOJ-1; carcinoma-associated protein HOJ-1 homolog; EGK_03473; HOJ1; HoJ-1; mHoj-1; Ras association (RalGDS/AF-6) domain family (N-terminal) member 8; Ras association (RalGDS/AF-6) domain family 8; Ras association domain family member 8; ras association domain-containing protein 8; RASSF8; RGD1308820
Common Name RASSF8
Gene Symbol RASSF8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TEFKQKVLNNCKTTADELKKLIRLQTEKLQSIEKQLESNEIEIRFWEQKYNSNLEEEIVRLEQKIKRNDVEIEEEEFWENELQIE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis