missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RASA3 (aa 736-811) Control Fragment Recombinant Protein

Product Code. 30180596
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30180596

Brand: Invitrogen™ RP98013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111244 (PA5-111244. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Human GAP1-InsP4 BP, also designated Ras p21 protein activator (GTPase-activating protein) 3 [Ins(1,3,4,5)P4-binding protein], is an 829-amino acid protein that binds phospholipids in both a calcium-dependent and -independent manner. GAP1, one of the Ras GTPase-activating protein families, comprises four distinct genes, including GAP1(m), GAP1-InsP4 BP, MRASAL (murine Ras GTPase-activating-like) and KIAA0538. This family contains an N-terminal tandem C2 domain, a GAP-related domain and a C-terminal pleckstrin homology (PH) domain. The PH domains of GAP1-InsP4 BP are essential for membrane targeting via binding of specific phospholipids. Following agonist-stimulated PtdIns(3,4,5)P(3) production, group I family PH domain containing proteins like GAP1-InsP4 BP specifically bind inositol phosphates, which are subsequently targeted to the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14644
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 22821
Name Human RASA3 (aa 736-811) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI326412; C86362; E130011G04; GAP1(IP4BP); GAP1-InsP4 BP; GAP1IP4BP; GAPIII; GAPIII activator 3; GTPase activating protein III; GTPase-activating protein 1 family, inositol 1,3,4,5-tetrakisphosphate-binding protein; ins P4-binding protein; Ins(1,3,4,5)P4-binding protein; ras GTPase-activating protein 3; Ras GTPase-activating protein III; RAS p21 protein activator 3; Rasa3; RP11-245B11.3; R-Ras gap; R-Ras GTP activating protein; R-ras GTPase activating protein; R-ras RTPase activating protein-3; scat
Common Name RASA3
Gene Symbol RASA3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LFNLYMSKLEKMQEACGSKSVYDGPEQEEYSTFVIDDPQETYKTLKQVIAGVGALEQEHAQYKRDKFKKTKYGSQE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.