missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RAPSN (aa 226-310) Control Fragment Recombinant Protein

Product Code. 30212425
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212425

Brand: Invitrogen™ RP97428

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58801 (PA5-58801. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of a family of proteins that are receptor associated proteins of the synapse. The encoded protein contains a conserved cAMP-dependent protein kinase phosphorylation site, and plays a critical role in clustering and anchoring nicotinic acetylcholine receptors at synaptic sites by linking the receptors to the underlying postsynaptic cytoskeleton, possibly by direct association with actin or spectrin. Mutations in this gene may play a role in postsynaptic congenital myasthenic syndromes. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13702
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5913
Name Human RAPSN (aa 226-310) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 43 kD receptor-associated protein of the synapse; 43 kda postsynaptic protein; 43 kDa receptor-associated protein of the synapse; 43 kDa; 43 kDa acetylcholine receptor-associated protein; acetylcholine receptor-associated 43 kda protein; AChR associated protein at synapses; acetylcholine receptor associated protein (at synapses; 43 kDa protein; likely to cluster nicotinic AChRs; CMS11; CMS1D; CMS1E; CMS4C; FADS; MGC3597; Nraps; Raps; Rapsn; rapsyn; receptor associated protein of the synapse; receptor-associated protein of the synapse; receptor-associated protein of the synapse, 43 kDa; receptor-associated protein of the synapse, 43 kD; RING finger protein 205; RNF205; twitch once; two)
Common Name RAPSN
Gene Symbol RAPSN
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MECCEESMKIALQHGDRPLQALCLLCFADIHRSRGDLETAFPRYDSAMSIMTEIGNRLGQVQALLGVAKCWVARKALDKALDAIE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.