missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RALGAPB (aa 1155-1258) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62410 (PA5-62410. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Non-catalytic subunit of the heterodimeric RalGAP1 and RalGAP2 complexes which act as GTPase activators for the Ras-like small GTPases RALA and RALB.
Specifications
Specifications
| Accession Number | Q86X10 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 57148 |
| Name | Human RALGAPB (aa 1155-1258) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 9330195P16; AI507211; B230339M05Rik; DKFZp781M2411; KIAA1219; mKIAA1219; p170; p170 {ECO:0000303; PubMed:19520869}; Ral GTPase activating protein non-catalytic beta subunit; Ral GTPase activating protein non-catalytic subunit beta; Ral GTPase activating protein, beta subunit (non-catalytic); ral GTPase-activating protein beta subunit; ral GTPase-activating protein subunit beta; ralgapb; ralgapb {ECO:0000303; RalGAPbeta; RGD1306861; zmp:0000000942 |
| Common Name | RALGAPB |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction