missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RAB25 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marca: R&D Systems AF6605
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Rab25 Polyclonal specifically detects Rab25 in Human samples. It is validated for Western Blot.
Especificaciones
| Rab25 | |
| Polyclonal | |
| Unconjugated | |
| P57735 | |
| RAB25 | |
| E. coli-derived recombinant human RAB25 Met1-Ala195 Accession # P57735 | |
| 100 μg | |
| Primary | |
| Detects human RAB25 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| CATX8, CATX-8RAB11C, RAB11C, RAB25, member RAS oncogene family, ras-related protein Rab-25 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 57111 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido