missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human R-Spondin 3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£153.00 - £408.00
Specifications
| Antigen | R-Spondin 3 |
|---|---|
| Clone | 1028220 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18743727
![]() |
R&D Systems
MAB3500-SP |
25 μg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18720277
![]() |
R&D Systems
MAB3500-100 |
100 μg |
£408.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
R-Spondin 3 Monoclonal specifically detects R-Spondin 3 in Human samples. It is validated for ELISA.Specifications
| R-Spondin 3 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| Cristin 1, CRISTIN1, FLJ14440, hPWTSR, hRspo3, Protein with TSP type-1 repeat, PWTSR, Roof plate-specific spondin-3, RSPO3, RSpondin 3, R-spondin 3 homolog (Xenopus laevis), R-spondin-3, Thrombospondin type-1 domain-containing protein 2, thrombospondin, type I, domain containing 2, THSD2 | |
| Chinese Hamster Ovary cell line CHO-derived human R-Spondin 3, Gln22-His272, Accession # Q9BXY4 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1028220 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BXY4 | |
| 84870 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title