missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human R-Spondin-2 Recombinant Protein, PeproTech®

Product Code. 17819932
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17819932

Brand: Gibco™ 120431MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human R-Spondin-2 is a 24.4 kDa protein consisting of 212 amino acid residues. Due to glycosylation, R-Spondin-2 migrates at an apparent molecular weight of approximately 30.0 kDa by SDS PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The R-Spondin (Rspo) proteins belong to the Rspo family of Wnt modulators. Currently, the family consists of four structurally-related, secreted ligands (Rspo 1-4), all containing furin-like and thrombospondin structural domains. The Rspo proteins can interact with the Frizzled/LRP6 receptor complex in a manner that causes the stabilization, and resulting accumulation, of the intracellular signaling protein, beta-catenin. This activity effectively activates and increases the subsequent nuclear signaling of beta-catenin. R-Spondin can also bind to the previously discovered G-protein coupled receptors, LGR-4 and LGR-5. Rspo/beta-catenin signaling can act as an inducer of the transformed phenotype, and can also regulate the proliferation and differentiation of certain stem cell populations.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6UXX9
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 340419
Molecular Weight (g/mol) 24.4 kDa
Name Human R-Spondin-2
Quantity 1 mg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 2610028F08Rik; AA673245; CRISTIN2; cristin-2; cysteine-rich and single thrombospondin domain-containing protein 2; D430027K22; ftls; hRspo2; mCristin-2; RGD1562331; Roof plate-specific spondin-2; Rspo2; R-spondin 2; R-spondin 2 homolog; R-spondin 2 homolog (Xenopus laevis); R-spondin-2; UNQ9384/PRO34209
Common Name RSPO2
Gene Symbol RSPO2
Biological Activity R-Spondin-2 enhances BMP-2-mediated differentiation of MC3T3-E1 cells. The expected ED50 for this effect is 0.8 -2.0 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence ASYVSNPICK GCLSCSKDNG CSRCQQKLFF FLRREGMRQY GECLHSCPSG YYGHRAPDMN RCARCRIENC DSCFSKDFCT KCKVGFYLHR GRCFDECPDG FAPLEETMEC VEGCEVGHWS EWGTCSRNNR TCGFKWGLET RTRQIVKKPV KDTILCPTIA ESRRCKMTMR HCPGGKRTPK AKEKRNKKKK RKLIERAQEQ HSVFLATDRA NQ
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.