missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PYK2 (aa 749-832) Control Fragment Recombinant Protein

Product Code. 30194772
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30194772

Brand: Invitrogen™ RP95203

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82904 (PA5-82904. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PTK2B (FAK2) is a cytoplasmic tyrosine kinase involved in calcium-induced regulation of ion channels and activation of the map kinase signaling pathway. PTK2B regulates reorganization of the actin cytoskeleton, cell polarization, cell migration, adhesion, spreading, and bone remodeling. FAK2 plays a role in the regulation of the humoral immune response and is required for normal levels of marginal B-cells in the spleen and normal migration of splenic B-cells. It functions in signaling downstream of integrin and collagen receptors, immune receptors, G-protein coupled receptors, cytokine, chemokine, and growth factor receptors, and mediates responses to cellular stress. Aberrant FAK2 expression may play a role in cancer cell proliferation, migration, and invasion in tumor formation and metastasis. Elevated expression has been seen in gliomas, hepatocellular carcinoma, lung cancer, and breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14289
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2185
Name Human PYK2 (aa 749-832) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CADTK; CAK beta; CAKB; CAKbeta; CAK-beta; calcium-dependent tyrosine kinase; Calcium-regulated non-receptor proline-rich tyrosine kinase; cell adhesion kinase beta; cellular adhesion kinase beta; E430023O05Rik; FADK; FADK 2; FADK2; FAK; FAK1; FAK2; focal adhesion kinase 2; FRNK; PKB; pp125FAK; proline-rich tyrosine kinase 2; protein kinase B; protein tyrosine kinase 2 beta; protein-tyrosine kinase 2-beta; PTK; PTK2 protein tyrosine kinase 2 beta; PTK2B; PTK2B protein tyrosine kinase 2 beta; PYK2; RAFTK; related adhesion focal tyrosine kinase
Common Name PYK2
Gene Symbol PTK2B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TLTSPMEYPSPVNSLHTPPLHRHNVFKRHSMREEDFIQPSSREEAQQLWEAEKVKMRQILDKQQKQMVEDYQWLRQEEKSLDPM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.