missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PU.1/Spi-1 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC5870N-100UG
This item is not returnable.
View return policy
Description
PU.1/Spi-1 Monoclonal specifically detects PU.1/Spi-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| PU.1/Spi-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_001074016 | |
| SPI1 | |
| E. coli-derived recombinant human PU.1/Spi-1 Met1-Lys169 Accession # NP_001074016 | |
| RUO | |
| 6688 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 732322 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| hematopoietic transcription factor PU.1, OF, PU.1, SFPI1, SFPI131 kDa transforming protein, SPI1, SPI-1, SPI-1 proto-oncogene, SPI-A, spleen focus forming virus (SFFV) proviral integration oncogene spi1,31 kDa-transforming protein, transcription factor PU.1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human PU.1/Spi-1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Spi-B, recombinant mouse (rm) PU.1/Spi-1, or rmSpi-B is observed. In Western blots, approximately 25% cross-reactivity with rhSpi-B and rmPU.1/Spi-1 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction