missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PTPRG (aa 37-135) Control Fragment Recombinant Protein

Product Code. 30200723
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200723

Brand: Invitrogen™ RP107321

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66673 (PA5-66673. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

RPTP gamma, also known as Receptor-type tyrosine-protein phosphatase gamma, R-PTP-gamma or PTPRG is a protein tyrosine phosphatase (PTP) is a candidate tumor suppressor gene since it is located on human chromosome 3p14.2-p21, a region frequently deleted in certain types of renal and lung carcinomas. In situ hybridization analysis reveals that RPTP gamma mRNA is expressed in specific regions of the brain and that the localization of RPTP gamma changes during brain development. RPTP gamma is composed of a putative extracellular domain, a single transmembrane domain, and a cytoplasmic portion with two tandem catalytic tyrosine phosphatase domains. The extracellular domain contains a stretch of 266 amino acids with striking homology to the zinc-containing enzyme carbonic anhydrase (CAH), indicating that RPTP gamma and RPTP beta (HPTP zeta) represent a subfamily of receptor tyrosine phosphatases. RPTP gamma may have a function other than catalysis of hydration of metabolic CO2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P23470
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5793
Name Human PTPRG (aa 37-135) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5430405N12Rik; AW046354; AW549872; H_RG317H01.1; HPTPG; protein tyrosine phosphatase gamma; protein tyrosine phosphatase, receptor type G; protein tyrosine phosphatase, receptor type, G; protein tyrosine phosphatase, receptor type, gamma polypeptide; protein-tyrosine phosphatase gamma; Protein-tyrosine phosphatase gamma precursor (R-PTP-gamma); Ptpg; Ptprg; receptor type protein tyrosine phosphatase gamma; receptor tyrosine phosphatase gamma; receptor-like protein tyrosine phosphatase gamma; receptor-type protein phosphatase gamma; receptor-type tyrosine-protein phosphatase gamma; RPTP gamma A; RPTP gamma B; RPTP gamma C; RPTP gamma S; RPTPG; RPTPgamma; R-PTP-GAMMA
Common Name PTPRG
Gene Symbol PTPRG
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ALHENRHGSAVQIRRRKASGDPYWAYSGAYGPEHWVTSSVSCGGRHQSPIDILDQYARVGEEYQELQLDGFDNESSNKTWMKNTGKTVAILLKDDYFVS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.