missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PTPN21 (aa 187-267) Control Fragment Recombinant Protein

Product Code. 30207139
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30207139

Brand: Invitrogen™ RP108627

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84944 (PA5-84944. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein tyrosine phosphatase (PTP) family of proteins are signaling molecules that regulate processes such as cell growth, cell differentiation, oncogenic transformation and the mitotic cycle. PTPN21 (protein tyrosine phosphatase, non-receptor type 21), also known as PTPD1 or PTPRL10, is a 1,174 amino acid member of the PTP family and localizes to both the cytoplasm and the cytoskeleton. Containing one FERM domain and one tyrosine-protein phosphatase domain, PTPN21 functions to catalyze the water-dependent conversion of a protein tyrosine phosphate into a protein tyrosine and a free phosphate and, via this catalytic activity, may be involved in liver regeneration and spermatogenesis. The gene encoding PTPN21 maps to human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16825
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11099
Name Human PTPN21 (aa 187-267) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias dJ1175B15.2 (protein tyrosine phosphatase, non-receptor type 21); protein tyrosine phosphatase 2 E; protein tyrosine phosphatase, non-receptor type 21; protein-tyrosine phosphatase 2 E; Protein-tyrosine phosphatase D1; protein-tyrosine phosphatase PTP-RL10; Ptp2E; PTPD1; Ptpn21; PTPRL10; Tyrosine-protein phosphatase non-receptor type 21
Common Name PTPN21
Gene Symbol PTPN21
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KVALLHQKYRGLTAPDAEMLYMQEVERMDGYGEESYPAKDSQGSDISIGACLEGIFVKHKNGRHPVVFRWHDIANMSHNKS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.