missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human PTPN20A (aa 60-87) Control Fragment Recombinant Protein

Produktkode 30200955
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
Förpackningsstorlek
100µL
Denne vare kan ikke returneres. Se returpolitik

Produktkode 30200955

missing translation for 'mfr': Invitrogen™ RP104358

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (36%), Rat (36%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60332 (PA5-60332. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Tyrosine-protein phosphatase targeted to sites of actin polymerization in response of varied extracellular stimuli. Has tyrosine phosphatase activity towards various tyrosyl phosphorylated substrates.
TRUSTED_SUSTAINABILITY

Tekniske data

Accession Number Q4JDL3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 26095
Name Human PTPN20A (aa 60-87) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias bA142I17.1; bA42B19.1; CT126; hPTPN20; protein tyrosine phosphatase, non-receptor type 20; protein tyrosine phosphatase, non-receptor type 20 A; protein tyrosine phosphatase, non-receptor type 20 B; PTPN20; PTPN20A; PTPN20B; Tyrosine-protein phosphatase non-receptor type 20
Common Name PTPN20A
Gene Symbol PTPN20
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NFPHNDYEDVFEEPSESGSDPSMWTARG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.