missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human PTHrP Recombinant Protein, PeproTech®

Product Code. 17820553
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17820553

Brand: Gibco™ 1000910UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human PTHrP is a 9.8 kDa linear polypeptide of 86 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the parathyroid hormone family. This hormone regulates endochondral bone development and epithelial-mesenchymal interactions during the formation of the mammary glands and teeth. This hormone is involved in lactation possibly by regulating the mobilization and transfer of calcium to the milk. The receptor of this hormone, PTHR1, is responsible for most cases of humoral hypercalcemia of malignancy. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed. There is also evidence for alternative translation initiation from non-AUG (CUG and GUG) start sites, in-frame and downstream of the initiator AUG codon, to give rise to nuclear forms of this hormone.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12272
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 5744
Molecular Weight (g/mol) 9.8 kDa
Name Human PTHrP
Quantity 10 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias BDE2; calciotropic hormone, growth factor; HHM; osteostatin; parathyroid hormone like hormone; parathyroid hormone related peptide; parathyroid hormone-like hormone; parathyroid hormone-like hormone preproprotein; parathyroid hormone-like peptide; parathyroid hormone-like protein; parathyroid hormone-like related protein; parathyroid hormone-related peptide; parathyroid hormone-related protein; parathyroid-like peptide; PLP; PTHLH; PTH-like; PTHR; PTH-related peptide; PTH-related protein; Pthrp; PTH-rP; PTHrP[107-139 ]; PTHrP[1-36 ]; PTHrP[38-94 ]
Common Name PTHLH
Gene Symbol PTHLH
Conjugate Unconjugated
Recombinant Recombinant
Sequence AVSEHQLLHD KGKSIQDLRR RFFLHHLIAE IHTAEIRATS EVSPNSKPSP NTKNHPVRFG SDDEGRYLTQ ETNKVETYKE QPLKTP
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.