missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PTH1R/PTHR1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5709
This item is not returnable.
View return policy
Description
PTH1R/PTHR1 Polyclonal specifically detects PTH1R/PTHR1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PTH1R/PTHR1 | |
| Polyclonal | |
| Unconjugated | |
| Q03431 | |
| PTH1R | |
| Chinese hamster ovary cell line CHO-derived recombinant human PTH1 R Tyr23-Met189 Accession # Q03431 | |
| 100 μg | |
| Primary | |
| Detects human PTH1 R in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| MGC138426, parathyroid hormone 1 receptorMGC138452, parathyroid hormone/parathyroid hormone-related peptide receptor, parathyroid hormone/parathyroid hormone-related protein receptor, PFE, PTH/PTHr receptor, PTH/PTHrP type I receptor, PTH1 receptor, PTHR, PTHR1, PTHRPTHR1parathyroid hormone receptor 1, seven transmembrane helix receptor | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5745 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction