missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PTH Recombinant Protein

Product Code. 11524950
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11524950 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11524950 Supplier Invitrogen™ Supplier No. RP10937

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This is a recombinant protein suitable for use as a control in ELISA and Western blot applications. Reconstitute the lyophilized product in sterile water not less than 100 μg/mL.

This gene encodes a member of the parathyroid family of proteins. The encoded preproprotein is proteolytically processed to generate a protein that binds to the parathyroid hormone/parathyroid hormone-related peptide receptor and regulates blood calcium and phosphate levels. Excess production of the encoded protein, known as hyperparathyroidism, can result in hypercalcemia and kidney stones. On the other hand, defective processing of the encoded protein may lead to hypoparathyroidism, which can result in hypocalcemia and numbness.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01270
For Use With (Application) Control
Formulation 1.15 mg sodium citrate, citric acid with 0.1117 mg EDTA, 0.2 mg Tween 80, 50 mg mannitol, 7.31 mg NaCl and no preservative
Gene ID (Entrez) 5741
Molecular Weight (g/mol) 4.1 kDa
Name Human PTH
Quantity 500 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias hypothalamic parathyroid hormone; parathormone; Parathyrin; parathyroid hormone; parathyroid hormone 1; preproparathyroid hormone; prepro-PTH; PTH; PTH-(1-84); Pth1; Pthp; Pthr1
Common Name PTH
Gene Symbol PTH
Biological Activity Specific Activity = 10,000 U/mg; determined by the UMR106 cell/cAMP method.
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Sequence Human PTH recombinant protein contains 34 amino acids
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Lyophilized
Protein Subtype Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.