missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PTGFRN (aa 367-471) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53547 (PA5-53547. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Inhibits the binding of prostaglandin F2-alpha (PGF2-alpha) to its specific FP receptor, by decreasing the receptor number rather than the affinity constant. Functional coupling with the prostaglandin F2-alpha receptor seems to occur. {ECO:0000250}.
Specifications
Specifications
| Accession Number | Q9P2B2 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 5738 |
| Name | Human PTGFRN (aa 367-471) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 4833445A08Rik; AU042434; CD315; CD9 partner 1; CD9P1; CD9P-1; EWIF; EWI-F; FPRP; Glu-Trp-Ile EWI motif-containing protein F; KIAA1436; prostaglandin F2 receptor inhibitor; prostaglandin F2 receptor negative regulator; Prostaglandin F2-alpha receptor regulatory protein; prostaglandin F2-alpha receptor-associated protein; Ptgfrn; SMAP-6; Trim45 |
| Common Name | PTGFRN |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction