missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PSMD8 (aa 203-345) Control Fragment Recombinant Protein

Product Code. 30209432
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209432

Brand: Invitrogen™ RP89240

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52329 (PA5-52329. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a non-ATPase subunit of the 19S regulator. A pseudogene has been identified on chromosome 1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48556
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5714
Name Human PSMD8 (aa 203-345) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 26 S proteasome non-ATPase regulatory subunit 8; 26 S proteasome regulatory subunit p31; 26 S proteasome regulatory subunit RPN12; 26 S proteasome regulatory subunit S14; 6720456J22Rik; AA407360; AL033291; AL033322; AL033323; C76433; epididymis secretory sperm binding protein Li 91 n; HEL-S-91 n; HIP6; HYPF; Nin1p; p31; proteasome (prosome, macropain) 26 S subunit, non-ATPase, 8; proteasome 26 S non-ATPase subunit 8; proteasome 26 S subunit, non-ATPase 8; proteasome 26 S subunit, non-ATPase, 8; Psmd8; Rpn12; S14
Common Name PSMD8
Gene Symbol PSMD8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NRVAEFHTELERLPAKDIQTNVYIKHPVSLEQYLMEGSYNKVFLAKGNIPAESYTFFIDILLDTIRDEIAGCIEKAYEKILFTEATRILFFNTPKKMTDYAKKRGWVLGPNNYYSFASQQQKPEDTTIPSTELAKQVIEYARQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.