missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PSMD12 (aa 263-361) Control Fragment Recombinant Protein

Product Code. 30196883
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30196883 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30196883 Supplier Invitrogen™ Supplier No. RP93548

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54701 (PA5-54701. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This protein is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that binds directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein is found in many histone deacetylase complexes, including mSin3 co-repressor complex. It is also present in protein complexes involved in chromatin assembly. This protein can interact with BRCA1 tumor-suppressor gene and may have a role in the regulation of cell proliferation and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00232
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5718
Name Human PSMD12 (aa 263-361) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1500002F15Rik; 26 S proteasome non-ATPase regulatory subunit 12; 26 S proteasome non-ATPase regulatory subunit 1-like protein; 26 S proteasome regulatory subunit p55; 26 S proteasome regulatory subunit RPN5; AI480719; fc34f08; MGC75406; P55; proteasome (prosome, macropain) 26 S subunit, non-ATPase, 12; proteasome 26 S non-ATPase subunit 12; proteasome 26 S subunit non-ATPase 12; proteasome 26 S subunit, non-ATPase 12; PSMD12; psmd12 protein; Rpn5; similar to proteasome 26 S non-ATPase subunit 12 isoform 2; wu:fb38a06; wu:fc34f08; wu:fc49f02
Common Name PSMD12
Gene Symbol PSMD12
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence WQQALKSVVLYVILAPFDNEQSDLVHRISGDKKLEEIPKYKDLLKLFTTMELMRWSTLVEDYGMELRKGSLESPATDVFGSTEEGEKRWKDLKNRVVEH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.