missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PSMB4 (aa 116-257) Control Fragment Recombinant Protein

Product Code. 30195109
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195109

Brand: Invitrogen™ RP89179

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52328 (PA5-52328. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P28070
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5692
Name Human PSMB4 (aa 116-257) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 26 kDa prosomal protein; HN3; HsBPROS26; HsN3; Lmp3; Low molecular mass protein 3; macropain beta chain; Multicatalytic endopeptidase complex beta chain; PROS26; PROS-26; Pros-27; proteasome (prosome, macropain) subunit, beta type 4; proteasome (prosome, macropain) subunit, beta type, 4; proteasome beta 4 subunit; proteasome beta chain; Proteasome chain 3; proteasome subunit beta 4; proteasome subunit beta type-4; proteasome subunit HsN3; proteasome subunit, beta type, 4; Psmb4; RN3; testis tissue sperm-binding protein Li 79 P
Common Name PSMB4
Gene Symbol PSMB4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VIDEELLGDGHSYSPRAIHSWLTRAMYSRRSKMNPLWNTMVIGGYADGESFLGYVDMLGVAYEAPSLATGYGAYLAQPLLREVLEKQPVLSQTEARDLVERCMRVLYYRDARSYNRFQIATVTEKGVEIEGPLSTETNWDIA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.