missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PSMB10/MECL1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF7684
This item is not returnable.
View return policy
Description
PSMB10/MECL1 Polyclonal specifically detects PSMB10/MECL1 in Human samples. It is validated for Western Blot.
Specifications
| PSMB10/MECL1 | |
| Polyclonal | |
| Unconjugated | |
| P40306 | |
| PSMB10 | |
| E. coli-derived recombinant human PSMB10/MECL1 Thr40-Glu273 Accession # P40306 | |
| 100 μg | |
| Primary | |
| Detects human PSMB10/MECL1 in direct ELISAs and Western blots. In direct ELISAs, less than 6% cross-reactivity with recombinant human PSMB7 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| beta2i, FLJ00366, LMP10, LMP10Proteasome subunit beta-2i, Low molecular mass protein 10, Macropain subunit MECl-1, MECl-1, MECL1, MECL1EC 3.4.25.1, MGC1665, Multicatalytic endopeptidase complex subunit MECl-1, proteasome (prosome, macropain) subunit, beta type, 10, proteasome catalytic subunit 2i, Proteasome MECl-1, proteasome subunit beta 7i, proteasome subunit beta type-10, Proteasome subunit beta-2i, proteasome subunit MECL1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5699 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction