missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PSGR Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB10669-100
This item is not returnable.
View return policy
Description
PSGR Monoclonal antibody specifically detects PSGR in Human samples. It is validated for Immunohistochemistry
Specifications
| PSGR | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| HPRAJ, Olfactory Receptor OR11-16, olfactory receptor, family 51, subfamily E, member 2, olfactory receptor, family 51, subfamily E, member 3 pseudogene, olfactory receptor, family 52, subfamily A, member 2, OR51E2, OR51E3P, OR52A2, prostate specific G-protein coupled receptor, Prostate-specific G-protein coupled receptor, PSGRolfactory receptor 51E2 | |
| Human PSGR synthetic peptide, Accession # Q9H255 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 1033009 | |
| Immunohistochemistry 5-25 ug/mL | |
| Q9H255 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 81285 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction