missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PSGL-1/CD162 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB996R-100UG
This item is not returnable.
View return policy
Description
PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Human samples. It is validated for Flow Cytometry.
Specifications
| PSGL-1/CD162 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA, SELPLG | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human PSGL-1/CD162. Recognizes sLe x -bearing core 2 O-glycan stuctures. It does not recognize sLe x on an extended core 1 O-glycan. The sLe x -bearing, core 2 O-glycan structure decorates the P-Selectin ligand PSGL-1, and the presence of this glycan structure is required for high affinity P-Selectin binding (1). This antibody stains human and canine leukocytes but does not recognize monkey, mouse, rabbit, porcine, feline or bovine leukocytes. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| CHO131 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| SELPLG | |
| CHO Chinese hamster ovary cell line transfected with human PSGL-1/CD162 | |
| RUO | |
| 6404 | |
| Human | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction