missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PSGL-1/CD162 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9962T-100UG
This item is not returnable.
View return policy
Description
PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Human samples. It is validated for Flow Cytometry.
Specifications
| PSGL-1/CD162 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA, SELPLG | |
| Chinese hamster ovary cell line CHO-derived recombinant human PSGL-1/CD162, Gln42-Gly295, Accession # Q14242 | |
| 100 μg | |
| 6404 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 688124 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q14242 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human PSGL-1/CD162 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction