missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PSENEN Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6859
This item is not returnable.
View return policy
Description
PSENEN/PEN2 Monoclonal specifically detects PSENEN/PEN2 in Human samples. It is validated for Immunohistochemistry.
Specifications
| PSENEN/PEN2 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q9NZ42 | |
| PSENEN | |
| Peptide containing human PSENEN Gly89-Gly98 Accession # Q9NZ42 | |
| 100 μg | |
| Primary | |
| Detects human PSENEN in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 665521 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| gamma-secretase subunit PEN-2, MSTP064, PEN-2, PEN2hematopoietic stem/progenitor cells protein MDS033, presenilin enhancer 2 homolog (C. elegans), Presenilin enhancer protein 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 55851 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction