missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PRUNE2 (aa 97-178) Control Fragment Recombinant Protein

Product Code. 30210449
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210449

Brand: Invitrogen™ RP93019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54592 (PA5-54592. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PRUNE2 gene, also named BMCC1, encodes protein prune homolog 2. PRUNE2 gene are overlapping with PCA3, one of the most prostate cancer specific markers. PRUNE2 may play an important role in regulating differentiation, survival and aggressiveness of the tumor cells. Besides its high level of expression seen in the nervous system (brain, cerebellum and spinal cord), it is also expressed at high levels in noneuroblastoma, rhabdomyosarcoma, melanoma and some osteosarcoma cell lines, whereas at only low levels in cancer cell lines of liver, breast, thyroid and colon. The Calculated molecular weight of this protein is 340 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8WUY3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 158471
Name Human PRUNE2 (aa 97-178) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6330414G02Rik; A214N16.3; A230083H22Rik; A330102H22Rik; bA214N16.3; BCH motif-containing molecule at the carboxyl terminal region 1; Bmcc1; BNIP2 motif containing molecule at the carboxyl terminal region 1; BNIP2 motif-containing molecule at the C-terminal region 1; BNIPXL; C9orf65; DKFZp762K117; KIAA0367; mKIAA0367; neuronal protein; olfaxin; protein prune homolog 2; prune homolog 2; prune homolog 2 (Drosophila); PRUNE2; RGD1311350; RP11-214N16.3; RP11-58J3.2; truncated PRUNE2
Common Name PRUNE2
Gene Symbol PRUNE2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LNDEGKLSITLVGSSVLASEDKTLESAVVKVINPVEQSDANVEFRESSSSLVLKEILQEAPELITEQLAHRLRGSILFKWMT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.