missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PRRX1 (aa 152-211) Control Fragment Recombinant Protein

Product Code. 30196484
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196484

Brand: Invitrogen™ RP108489

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111567 (PA5-111567. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The homeobox DNA-binding domain is a 60 amino acid motif that is conserved among many species and functions to bind DNA via a helix-turn-helix structure, thereby playing a role in transcriptional regulation and the control of gene expression. PRX1 (paired related homeobox 1), also known as PRRX1, PMX1 or PHOX1, is a 245 amino acid protein that contains one OAR domain and one homeobox DNA-binding domain and belongs to the paired homeobox family. Localized to the nucleus, PRX1 functions as a transcriptional co-activator that enhances the DNA-binding activity of serum response factor (SRF), thereby mediating the induction of SRF-dependent gene expression by growth and differentiation factors. Additionally, PRX1 regulates the transcriptional activities of creatine kinase-M (muscle), thereby playing a role in the establishment of mesodermal muscle types. PRX1 exists as two alternatively spliced isoforms, designated PMX1-A and PMX1-B.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P54821
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5396
Name Human PRRX1 (aa 152-211) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A230024N07Rik; AA755424; AGOTC; AI385634; AI843499; Homeobox protein K-2; homeobox protein mHox; Homeobox protein PHO x 1; K-2; mHox; paired mesoderm homeobox 1; paired mesoderm homeobox 1 isoform pmx-1 b; paired mesoderm homeobox protein 1; paired related homeobox 1; paired-related homeobox protein 1; PHO x 1; Pmx; Pm x 1; Prr x 1; Pr x 1; PRX-1; rHox
Common Name PRRX1
Gene Symbol PRRX1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NERAMLANKNASLLKSYSGDVTAVEQPIVPRPAPRPTDYLSWGTASPYRSSSLPRCCLHE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.