Learn More
Human Prox1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand:  R&D Systems AF2727
Product Code. 18080632
  • £516.00 / 100µg
Description
Prox1 Polyclonal specifically detects Prox1 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Prox1
Polyclonal
Unconjugated
Q92786
PROX1
E. coli-derived recombinant human Prox1 Pro2-Gln259 Accession # Q92786
100 μg
Primary
Detects human Prox1 in direct ELISAs and Western blots.
Human
IgG
Western Blot, Immunocytochemistry
LYOPH
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 5-15 ug/mL
Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1
Goat
Affinity Purified
RUO
5629
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.