missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Protocadherin gamma C3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £362.00
Specifications
| Antigen | Protocadherin gamma C3 |
|---|---|
| Clone | 926525 |
| Dilution | Western Blot 2 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18787188
![]() |
R&D Systems
MAB8364-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18708709
![]() |
R&D Systems
MAB8364 |
100 μg |
£362.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protocadherin gamma C3 Monoclonal specifically detects Protocadherin gamma C3 in Human samples. It is validated for Western Blot.Specifications
| Protocadherin gamma C3 | |
| Western Blot 2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cadherin-like 2, PC43, PC-43, PCDH2, PCDH-GAMMA-C3, PCDHGC3, protocadherin 2, protocadherin 43, protocadherin gamma subfamily C, 3, protocadherin gamma-C3, protocadherin-2, Protocadherin-43 | |
| PCDHGC3 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 926525 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UN70 | |
| 5098 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Protocadherin gamma C3 Met1-Tyr693 Accession # Q9UN70 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Protocadherin gamma C3 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title