missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Protocadherin-15 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6729-SP
This item is not returnable.
View return policy
Description
Protocadherin-15 Monoclonal specifically detects Protocadherin-15 in Human samples. It is validated for Immunocytochemistry.
Specifications
| Protocadherin-15 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| Q96QU1 | |
| PCDH15 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Protocadherin-15 Tyr28-Ala1376 Accession # Q96QU1 | |
| 25 μg | |
| Primary | |
| Detects human Protocadherin-15 in direct ELISAs. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 740523 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| autosomal recessive 23, cadherin-related family member 15, CDHR15, DFNB23, DKFZp667A1711, PCDH15, protocadherin-15, Protocadherin15, protocadherin-related 15, USH1F | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 65217 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction