missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Protein O-Glucosyltransferase 1/POGLUT1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6437
This item is not returnable.
View return policy
Description
Protein O-Glucosyltransferase 1/POGLUT1/KTELC1 Monoclonal specifically detects Protein O-Glucosyltransferase 1/POGLUT1/KTELC1 in Human samples. It is validated for Western Blot.
Specifications
| Protein O-Glucosyltransferase 1/POGLUT1/KTELC1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q8NBL1 | |
| POGLUT1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Protein O-Glucosyltransferase 1/POGLUT1 Arg24-Leu388 Accession # Q8NBL1 | |
| 100 μg | |
| Primary | |
| Detects human Protein O-Glucosyltransferase 1/POGLUT1 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 737509 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 9630046K23Rik, C3orf9, CAP10-like protein, 46 kDa, chromosome 3 open reading frame 9, CLP46, EC 2.4.1.-, hCLP46CAP10-like 46 kDa protein, KDELC family like 1, KDELCL1, KTEL (Lys-Tyr-Glu-Leu) containing 1, KTEL motif-containing protein 1, KTELC1, MDS010, MDSRP, MDSRPKTELC1, MGC32995, Myelodysplastic syndromes relative protein, POGLUT1, protein O-glucosyltransferase 1, Rumi, x 010 protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56983 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction